missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic-2,3-d2 Hydrazide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Maleic Hydrazide D2 |
| CAS | 2398483-97-9 |
| Molecular Formula | C4 D2 H2 N2 O2 |
| Quantity | 0.1 g |
| Synonym | Maleic Hydrazide D2, 1,2-Dihydro-3,6-pyridazinedione-4,5-d2, 3,6-Dihydroxypyridazine-4,5-d2, Maleic-d2 acid cyclic hydrazide, Maleic-d2 hydrazide |
| InChI Formula | InChI=1 S/C4H4N2O2/c7-3-1-2-4(8)6-5-3/h1-2 H,(H,5,7)(H,6,8)/i1D,2 D |
| SMILES | [2 H]C1=C([2 H])C(=O)NNC1=O |
| IUPAC Name | 4,5-dideuterio-1,2-dihydropyridazine-3,6-dione |
| Molecular Weight (g/mol) | 114.0991 |
| Formula Weight | 114.0398 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction