Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Product Code. 10409444 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Product Type:
GC Autosampler Priming Syringe, Luer-Lok
GC Autosampler Syringe
Gas Tight GC Autosampler Syringe, Luer-Lok
Unit Size:
Each
Product Code. 10409444
Brand: Thermo Scientific™ 365PL571
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Tight GC Autosampler Syringe, Luer-Lok
Volume (Metric) 25 mL
For Use With (Equipment) Gas Chromatography Systems
Replacement Needle 365RNL22