Learn More
Colder Products Company Male Luer Genderless MicroCNX Connector
Product Code. 30968813
Click to view available options
:
Connector Type: Male
Product Code. 30968813
Brand: Colder Products Company CNX170LMHT
MicroCNX™ Connectors are the only aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small-format biomanufacturing assemblies.
  • The MicroCNX line of connectors is engineered specifically for the challenging conditions of biological media transfer in bioprocessing, cell therapy, and gene therapy applications.
  • The connector comes in a range of tubing sizes; a luer connection option to create a sterile, closed connection on an existing luer connection; and the ULT version is ideal for assemblies for cryogenic freezing cassettes or connection to PVC tubing.
  • Finish: Molded White
  • Pressure Range: Up to 43.5 psi, 3 bar
  • Free Floating
  • Temperature Range: 34°F to 104°F (1°C to 40°C)
  • Seal Options: Silicone, platinum-cured, USP Class VI, ADCF
  • Non-Valved
Specifications
Autoclavable Autoclavable
Material Polycarbonate
Sterility Non-sterile
Color White, Blue
Product Type Connector