Learn More
3M™ Maintenance Sorbent Roll
Product Code. 17158600
Click to view available options
Width (Metric):
400 mm
960 mm
Unit Size:
1 roll
Product Code. 17158600
Brand: 3M™ 7000051880
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A scrim on one side increases the strength and helps to eliminate linting. Available in convenient widths that are perfect for lining drawers, covering workbenches, or masking larger areas from mist and overspray.
  • Designed for use in and around the work area to catch leaks and drips
  • For use on most non-aggressive liquids
  • Very easy to use and to retrieve
  • Designed to cover large surfaces
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Capacity (English) 85.3 gal.
Capacity (Metric) 322.9 L
Type Sorbent Roll
For Use With (Application) Workplace safety
Model M-B2002
Dimensions (L x W) 45.7 x 0.96 m
Length (English) 150 ft.
Length (Metric) 45.7 m
Width (English) 38 in.
Width (Metric) 960 mm