Learn More
3M™ Maintenance Sorbent Mini-Booms
Click to view available options
Length (Metric):
240 cm
Unit Size:
Pack of 6
Specifications
Specifications
| Type | Maintenance Sorbent Mini-Boom |
| Packaging Quantity | 6/Cs. |
| Length (Metric) | 240 cm |
| Weight (Metric) | 5.5 kg |