Learn More
Gilson™ Maintenance Kit for Pipetman™
Manufactured by Gilson™. Gilson™ Maintenance Kit for Pipetman™ are guaranteed to deliver value and assure reliability and precision.
Brand: Gilson™ F144501
This item is not returnable.
View return policy
Description
- Maintenance kit
Specifications
| For Pipetman: P2/P2G | |
| 1 Tip-holder, 3 piston seals and 3 O-rings |