Learn More
BOLA Magnetic Stirring Bar Retrievers
Product Code. 10288870
Click to view available options
Length (Metric):
200 mm
Unit Size:
Each
Product Code. 10288870
Brand: BOLA C37204
Only null left
Add to basket
Add to basket
Magnetic Stirring Bar Retrievers PTFE, with permanent magnet
PTFE-encapsulated stirring bar retriever with strong permanent magnet (Alnico 5), universal chemical resistance.
Specifications
Autoclavable Autoclavable
Color White
Diameter (Metric) 8 mm
Length (Metric) 200 mm