Learn More
Phenomenex™ Magnetic Screw Caps, 18 mm, with Septa, No Slit
Description
- Technique: Accessory
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Outer | 18 mm |
| Pre-slit | No |
| Sterility | Sterile |
| Septum | PTFE/Silicone |
| Closure Size | 18 mm |
| Marking Spot | No |
| Autoclavable | Autoclavable |
| Product Type | Cap |
| Septum Color | White/Blue |