Learn More
Brady™ Magnetic Plastic ADR Transport Signs
Product Code. 11790244
Click to view available options
Displayed Text:
1
1.4 1
1.5 1
1.6 1
3
4
Unit Size:
Each
Product Code. 11790244
Brand: Brady™ 223605
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material Magnetic Plastic
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 297 mm
Width (Metric) 297.00 mm
Shape Diamond
Displayed Text 4
Color White on Blue