Learn More
Brady™ Magnetic Plastic ADR Transport Signs
Product Code. 11760244
Click to view available options
Displayed Text:
1
1.4 1
1.5 1
1.6 1
3
4
Unit Size:
Each
Product Code. 11760244
Brand: Brady™ 223602
Specifications
Alternate Part Number ADR 3B-297*297-B0859
Material Magnetic Plastic
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 297 mm
Width (Metric) 297.00 mm
Shape Diamond
Displayed Text 3
Color White on Red