missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Magnetic Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
200 μg
60 μg
Unit Size:
1 set
Description
- Conjugates to specially treated magnetic particles
- No extensive optimization of the conjugation reaction
- Covalent attachment of antibodies to magnetic particles
- Bead size: 0.5μm
- Minimizes risk of unspecific binding
Applications: Immunoprecipitation experiments
Storage: Store at 4°C
Note: Kits are guaranteed for 6 months from date of receipt
Specifications
Specifications
| Quantity | 60 μg |
| Product Type | Magnetic antibody labeling kit |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction