Learn More
Brady™ Magnet Accessory for M210 and M211 Label Printers
Description
This magnet accessory allows you to attach your Brady™ M210 or M211 Label Printer to panels and other magnetic-receptive surfaces during use or storage.
- Attaches to back of printer with screws to ensure stability
- Lightweight and slim design does not hinder handheld use of the printer
- Printer sold separately
Specifications
Specifications
| Product Type | Magnetic Attachment |
| Color | Black |
| For Use With (Equipment) | Brady M210 Label Printer, BMP21-PLUS Label Printer, Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer |
| Width (Metric) | 63.50 mm |
| Height (English) | 2.5 in |
| Height (Metric) | 63.50 mm |
| Depth (English) | 0.5 in. |
| Depth (Metric) | 12.70 mm |
| Dimensions (L x W) | 63.50 mm (W) x 63.50 mm (H) x 12.70 mm (D) |
| Color Capability | Gray |
| Show More |