missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tert-butoxide, 93+%

Product Code. 10064343 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10064343

Brand: Thermo Scientific Acros 390472500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 32149-57-8 | C8H18MgO2 | 170.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32149-57-8
Molecular Formula C8H18MgO2
Molecular Weight (g/mol) 170.53
MDL Number MFCD03788761
InChI Key RTKCPZYOLXPARI-UHFFFAOYSA-N
Synonym magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
PubChem CID 12040442
IUPAC Name magnesium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]

Specifications

Color White to Gray
Assay Percent Range 93+%
Packaging Plastic bottle
Quantity 250 g
Solubility Information Solubility in water: reacts violently
Formula Weight 170.53
Percent Purity 93+%
Physical Form Powder
Chemical Name or Material Magnesium tert-butoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.