missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate concentrate, Honeywell Fluka™

Product Code. 15685230 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
This item is not returnable. View return policy

Product Code. 15685230

Brand: Honeywell-Fluka 381466X1EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

for 1L standard solution, 0.1 M MgSO4

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 6 x 1 Ea.
UN Number NONH for all modes of transport
IUPAC Name magnesium(2+) sulfate
Formula Weight 120.37g/mol
Chemical Name or Material Magnesium sulfate concentrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.