Learn More
Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation
Description
A useful laboratory reagent, and a good reference white color for colorimetry. The interaction of magnesium oxide substrates with nickel ferrite films has been investigated using a wide range of electron optical techniques. Considerable interdiffusion is observed, which is likely to alter the magnetic properties of these films. Epitaxial ferrite-spinel layer is formed on magnesium oxide substrate. Additionally, Magnesium oxide in nanoparticles dispersed in alcohol have been reported to inhibit hydrolytic and oxidative degradation of paper documents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 Ea. |
| Solubility Information | Soluble in 5 M HCl (0.1 M) at 20°C,and dilute acids. Insoluble in water,and ethanol. |
| Chemical Name or Material | Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.