missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxalate dihydrate, 98.5+%

Product Code. 11375028
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11375028

Brand: Thermo Scientific Alfa Aesar 043976.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-66-0 | C2H2MgO4+2 | 114.339 g/mol

Magnesium oxalate dihydrate is in the synthesis of nano sized magnesium oxide. It is also used as a catalyst, refractory materials, adsorbents, superconductors, and ferroelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-66-0
Molecular Formula C2H2MgO4+2
Molecular Weight (g/mol) 114.339
MDL Number MFCD00287279
InChI Key UHNWOJJPXCYKCG-UHFFFAOYSA-N
Synonym magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
PubChem CID 54611841
IUPAC Name magnesium;oxalic acid
SMILES C(=O)(C(=O)O)O.[Mg+2]

Specifications

Melting Point 150°C (decomposition)
Physical Form Powder, Pieces
Quantity 2 kg
UN Number UN3288
Merck Index 14,5676
Solubility Information Soluble in diluted mineral acid,water (0.038g/100g H2O ). Insoluble in organics solutions.
Formula Weight 148.36 (112.32 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Chemical Name or Material Magnesium oxalate dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.