missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium fluoride, 99% (metals basis excluding Ca & Na), Ca+Na <1%
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
Description
Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1261°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 99%, Ca+Na <1% (Metals basis excluding Ca and Na) |
| Solubility Information | Sparingly soluble in water. Slightly soluble in dilute HNO3Insoluble in ethanol. |
| Formula Weight | 62.3 |
| Odor | Odorless |
| Refractive Index | 1.35-1.38 |
| Density | 3.148 |
| Show More |
Safety and Handling
Product Identifier
- Magnesium fluoride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Magnesium fluoride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction