missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Magnesium Chloride Hexahydrate, MP Biomedicals™

Product Code. 11430532 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11430532

Brand: MP Biomedicals™ 02191421.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Can be generated from the acid leach solution of magnesite tailings. Magnesium Chloride Hexahydrate, MP Biomedicals has been used to prepare a buffer for the renaturation of denatured peptidoglycan hydrolytic enzymes. It is a potential phase change materi

Application include:

  • Potential phase change material (PCM) for storing thermal energy in solar cookers
  • To alter poly (vinyl alcohol) [PVA] films has been investigated
  • To reduce the flammability of cotton fabrics has been studied
  • To prepare renaturation buffer for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes
  • As a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones
TRUSTED_SUSTAINABILITY

Specifications

Grade ACS
Purity 99 to 102%
For Use With (Application) Magnesium chloride hexahydrate has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones.
Content And Storage Store at Room Temperature (15–30°C)
Quantity 100 g
Molecular Weight (g/mol) 203.3g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.