missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate hydroxide pentahydrate, light, 98%

Product Code. 15487515
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 15487515

Brand: Thermo Scientific Alfa Aesar A18070.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol

Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water and acids. Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56378-72-4
Molecular Formula C4H12Mg5O19
Molecular Weight (g/mol) 485.65
MDL Number MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Synonym acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Quantity 5000 g
Linear Formula 4MgCO3·Mg(OH)2·5H2O
Solubility Information Soluble in water and acids. Insoluble in alcohol.
Formula Weight 485.65 (395.58 Anhydrous)
Percent Purity 98%
Density 2.16
Chemical Name or Material Magnesium carbonate hydroxide pentahydrate, light

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.