Learn More
Brady Magicard™ - Monochrome Film - Gold
Click to view available options
Color:
Gold
Unit Size:
Each
Description
- This high-quality, monochrome ribbon guarantees to print 1000 single-sided cards.
- Includes a cleaning roller for dust free printing.
Specifications
Specifications
| Product Type | Printer Ribbon |
| Color | Gold |
| For Use With (Equipment) | Magicard 300, Magicard 600, Pronto 100 |
| Print Technology | Thermal Transfer |
| Length (English) | 32.8 ft. |
| Width (English) | 2.4 in. |
| Length (Metric) | 10 m |
| Width (Metric) | 60 mm |
| Dimensions (L x W) | 10 m x 60 mm |
| Description | Magicard Monochrome Film - Gold |
| Show More |