missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAGEB1 (Human) Recombinant Protein
Human MAGEB1 full-length ORF ( AAH13772, 1 a.a. - 347 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004112-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPRGQKSKLRAREKRRKAREETQGLKVAHATAAEKEECPSSSPVLGDTPTSSPAAGIPQKPQGAPPTTTAAAAVSCTESDEGAKCQGEENASFSQATTSTESSVKDPVAWEAGMLMHFILRKYKMREPIMKADMLKVVDEKYKEYFTEILNGASRRLELVFGLDLKEDNPSGHTYTLVSKLNLTNDGNLSNDWDFPRNGLLMPLLGVIFLKGNSATEEEIWKFMNVLGAYDGEEHLIYGEPRKFITQDLVQEKYLKYEQVPNSDPPRYQFLWGPRAYAETTKMKVLEFLAKMNGATPRDFPSHYEEALRDEEERAQVRSSVRAKRRTTATTFRARSRAPFSRSSHPM
Specifications
AAH13772 | |
melanoma antigen family B, 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAGEB1 | |
GST |
4112 | |
Wheat germ expression system | |
25 μg | |
DAM10, MAGE-Xp, MAGEL1, MGC9322 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |