Learn More
MP Biomedicals™ Magbeads FastRNA Kit for FFPE
Product Code. 17742448 Shop All MP Biomedical Products
Click to view available options
:
192 Preps
Product Code. 17742448
Brand: MP Biomedicals™ 116573192
MagBeads FastRNA Kit for FFPE is intended for rapid extraction of RNA from tissue, cells, blood, and other clinical samples. RNA can be used directly for RT-PCR, quantitative RT-PCR and so on.
MagBeads FastRNA Kit for FFPE is based on the purification method of high binding magnetic particles:
  • The sample is lysed and digested
  • RNA is released into the lysate
  • After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
  • The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
  • Finally the RNA was eluted with Elution Buffer
TRUSTED_SUSTAINABILITY
Specifications
Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type FFPE
Sequencing Type DNA and RNA Sequencing
No. of Reactions 192 Preps
compliance-icons compliance-icons
Product Identifier
  • Magbeads FastRNA Kit for FFPE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K