missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS MagBeads FastRNA Kit for Feces 5 Preps

Product Code. 31900319 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 Preps
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31900319 5 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31900319 Supplier MP BIOMEDICALS Supplier No. 116588000
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

High-yield RNA extraction from fecal samples, delivering up to 250 μg from 200 mg input. Streamlined workflow reduces handling steps, saving time and simplifying purification.

  • MagBeads FastRNA Kit for Feces is a scalable and automatable magnetic bead-based RNA purification technology designed for efficient isolation of total RNA from fecal samples. Ready-to-use versions are available (Cat. No. 117040300 & Cat. No. 117040400).
  • The composition of fecal samples largely depends on diet and may include fibers, undigested particles, bilirubin, complex polysaccharides, and lipids.
  • These compounds impair sample homogenization, reducing both the quantity and quality of the extracted RNA.
  • The MagBeads FastRNA Kit for Feces integrates state-of-the-art bead-beating technology, a lysis chemistry compatible with a wide range of samples, and proprietary inhibitor removal expertise.
  • A selective RNA-binding technology combined with FastDNase I enzyme treatment enables complete removal of DNA.
  • The kit streamlines RNA purification by overcoming challenges associated with diverse fecal sample types, making the process hassle-free and time-efficient.
  • A DNase I treatment step effectively removes contaminating DNA, ensuring high RNA yield and purity.
  • The extracted RNA is immediately compatible with downstream applications such as RT-qPCR amplification, eliminating the need for additional inhibitor or DNA removal steps.
  • Application Notes: The MagBeads FastRNA Kit for Feces is designed for rapid RNA isolation from fecal samples using magnetic bead automation technology.
  • Usage Statement: Research Use Only (RUO).
  • Key Applications: RT-PCR.

Specifications

Sample Type Feces
Product Type RNA Feces
Quantity 5 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.