Learn More
MP Biomedicals™ Magbeads FastDNA/RNA Kit for Virus
Product Code. 17722448 Shop All MP Biomedical Products
Click to view available options
:
192 Preps
Product Code. 17722448
Brand: MP Biomedicals™ 116571192
MagBeads FastDNA/RNA Kit for Virus is intended for extracting total viral nucleic acid from cell-free/low-cell content biological samples such as body fluids, serum, plasma, soaking solution, tissue homogenate, and culture supernatant.
MagBeads FastDNA/RNA Kit for Virus is based on the purification method of high binding magnetic particles:
  • The sample is lysed and digested with Protease
  • DNA/RNA is released into the lysate
  • After addition of magnetic particles and binding solution, DNA/RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
  • The adsorbed particles are washed with washing solution to remove proteins and impurities, washed with ethanol to remove salts, and finally DNA/RNA is eluted with Nuclease Free Water
TRUSTED_SUSTAINABILITY
Specifications
Format Kit
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Virus
Sequencing Type DNA and RNA Sequencing
No. of Reactions 192 Preps
compliance-icons compliance-icons
Product Identifier
  • Magbeads FastDNA/RNA Kit for virus
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K