Learn More
MP BIOMEDICALS Magbeads FastDNA Kit for Soil (Ready-to-Use for MPure-32) 96 Preps
Shop All MP Biomedical Products
Click to view available options
:
96 Preps
Description
- The MagBeads FastDNA Kit for Soil allows quick and efficient isolation of high-quality genomic DNA from soil in less than 60 mins.
- Samples are placed into Lysing Matrix E tube and used with FastPrep Instruments from MP Biomedicals to effectively lyse any cells included in environmental samples within 40 seconds:
- Bacteria
- Fungi
- Viruses
- Protists
- The kit is compatible with the MPure-32 automated nucleic acid extraction instrument.
- It consists of specially formulated reagents to eliminate :
- humic acid
- polysaccharides
- phenolic compounds
- and enzyme inhibitors from soil
- and thus allow extraction of highly pure genomic DNA ready for PCR, restriction digestion, electrophoresis, and other desired applications.
- Application Notes:
- Extraction of highly pure genomic DNA from different types of soil, including :
- organic soil, paddy soil
- flowerbed soil
- saline soil
- desert soil that is ready for downstream analyses such as:
- PCR
- restriction digestion
- electrophoresis
- and other desired applications.
- Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Includes | 96 Preps |
| Product Type | DNA Extraction |
Safety and Handling
Product Identifier
- (MPure-32) MagBeads FastDNA Kit for Soil
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Ethanol, Guanidine Hydrochloride, Isopropanol, Lithium Chloride, Disodium Dihydrogen Ethylenediamine