Learn More
MP Biomedicals™ Magbeads FastDNA Kit for Blood
Shop All MP Biomedical Products
Click to view available options
:
96 Preps
Description
MagBeads FastDNA Kit for Blood is based on the purification method of high binding magnetic particles:
- The sample is lysed and digested
- DNA is released into the lysate
- After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
- The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
- Finally the DNA was eluted with Elution Buffer
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Scale | Mini |
| Sample Type | Blood |
| Sequencing Type | DNA and RNA Sequencing |
| No. of Reactions | 96 Preps |
Safety and Handling
Product Identifier
- Magbeads FastDNA Kit for Blood
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Proteinase K