Learn More
Gilson™ MACROMAN™ Wall Mounts
Product Code. 15891727 Shop All Gilson Products
Click to view available options
Description:
Magnetic
Screw/Adhesive Tape
Unit Size:
Each
Product Code. 15891727
Brand: Gilson™ F107201
Only null left
Add to basket
Add to basket
These wall mounts provide safe and stable storage for your MACROMAN™ pipette controllers.
  • Keeps pipette controllers away from busy benches and desktops
  • Available in magnetic and screw varieties
  • Accessory for MACROMAN pipette controllers
  • Supplied individually
TRUSTED_SUSTAINABILITY
Specifications
Description Screw/Adhesive Tape
Color Black
Type Wall Mount
For Use With (Application) Storage
For Use With (Equipment) MACROMAN™ Pipette Controller