Learn More
Gilson™ MACROMAN™ Wall Mounts
Shop All Gilson Products
Click to view available options
Description:
Magnetic
Screw/Adhesive Tape
Unit Size:
Each
Description
- Keeps pipette controllers away from busy benches and desktops
- Available in magnetic and screw varieties
- Accessory for MACROMAN pipette controllers
- Supplied individually
Specifications
Specifications
| Description | Magnetic |
| Color | Black |
| Type | Wall Mount |
| For Use With (Application) | Storage |
| For Use With (Equipment) | MACROMAN™ Pipette Controller |