missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MACROMAN™ Side Stand for MACROMAN Pipette Controller

Product Code. 15841737 Shop All Gilson Products
Click to view available options
:
Side stand; accessory
This item is not returnable. View return policy

Product Code. 15841737

Brand: Gilson™ F107210

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This adjustable side stand is a replacement part for the MACROMAN™ pipette controller.

  • Integrated adjustable side stand
  • Unique feature for left- and right-handed users for a stable, safe and easy storage option on benchtops
  • Accessory for MACROMAN pipette controllers
  • Supplied individually
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Type Side Stand
For Use With (Application) Storage
For Use With (Equipment) MACROMAN™ Pipette Controller
Quantity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.