Learn More
Emma Safety Footwear Mace M Safety Shoes
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo / HRO
- Full grain leather
- Heat resistant outsole
- Hydro-Tec™ Sanitized Silver
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Mace M |
| Liner Material | Hydro-Tec™ Sanitized Silver |
| Material | Leather |
| Size | 48 |
| Toe Material | Steel |