Learn More
Tocris Bioscience™ M8 B hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
M8 B hydrochloride is a potent and selective TRPM8 channel blocker. Blocks cold- and icilin -induced TRPM8 activation in vitro (IC50 values are 7.8 and 26.9 nM respectively). Exhibits no effect at other TRP channels (IC50 > 20 μM). Reduces body temperature in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | M8 B hydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 432.96 |
| Inhibitors | M8 B hydrochloride |