Learn More
Brady™ M611 Label Printer
Product Code. 17865235 Shop All Brady Products
Change view
Click to view available options
Product Type:
Label Printer EU
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17865235 Label Printer EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17865235 Supplier Brady™ Supplier No. 317844
  • ONLINE EXCLUSIVE PRICE
    715.00€
    595.00€ / Pack of 1
    Save 120.00€
    16% Off
View all promotions
 Restricted Item
In Stock
Design, preview, and print any way you choose. The M611 Label Printer allows you to store more than 1,000 labels on board, access files easier and print multiple ways, on demand.
  • Print labels at 50.8 mm/sec. print speed
  • Design, preview and print any way you choose: from your phone, tablet, or computer
  • Store more than 1,000 labels on the printer, then print at the jobsite
  • Create and share label files to print from anywhere using Bluetooth or Wi-Fi
  • Features a compact and lightweight design
  • Allows connectivity for up to 5 users via Bluetooth
Specifications
Product Type Label Printer EU
Color Blue, Gray
Display Type Backlit Graphic Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 10.97 in.
Width (English) 4.46 in.
Length (Metric) 278.75 mm
Width (Metric) 113.28 mm
Height (English) 4.55 in.
Height (Metric) 115.66 mm
Color Capability Monocolor
Display Size 88.9 mm
Includes Printer, USB Cable, Quick Start Guide, AC Powercord (EU), USB with Product Info and Windows Driver, Brady Workstation Basic Design Suite, Roll of M6-11-427 and M61-R4310 Ribbon
Languages Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Greek, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Memory 256 MB Flash, 512 MB RAM
Model M611
Print Speed 50.8 mm/sec.
Resolution 300 dpi
Connector Types Bluetooth™ 5 Low Energy (Class II), USB 2.0, Wi-Fi
Weight (English) 3.52 lb.
Weight (Metric) 1.6 kg
Certifications/Compliance FCC Part 15 Class A
For Use With (Application) Asset Tracking, Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Safety Identification
Warranty 2 years
Power Supply Details AC Adaptor, Rechargeable Battery
Voltage 110/240 V
Alternate Part Number M611-EU-BT-W