Learn More
Brady™ M610 Label Printer
Product Code. 17855225
missing translation for 'orderingAttributeHoverText'
Keyboard Type:
AZERTY
QWERTY (EU)
missing translation for 'unitSize'
Each
Product Code. 17855225
missing translation for 'mfr': Brady™ 317792
Get the durability your work demands with the M610 Label Printer with enhanced capabilities like military grade shock resistance, below freezing operating temperatures, and the widest range of data entry options. Kit includes a hard case.
  • Design and print labels using a keypad or desktop software
  • Work longer with an enhanced Gen3 Li-ION battery
  • Print 4,500 labels on a full charge
  • Easily import and export label files, lists and graphics to the printer through USB 2.0 port (Type A and B) or USB flash drive
  • Print labels at 50.8 mm/sec. print speed
  • Get added durability with a printer that is resistant to military-grade shocks (MIL-STD-810G) and 1.8 m drops
  • Operate your printer in -18°C to +46°C temperatures
Specifications
Product Type Printer
Color Yellow, Gray
Display Type Backlit Graphic Color LCD
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 4.19 in.
Width (English) 4.22 in.
Length (Metric) 106.42 mm
Width (Metric) 107.19 mm
Height (English) 12.93 in.
Height (Metric) 328.42 mm
Color Capability Monocolor
Display Size 71.12 mm
Includes Printer, Hard Case, USB Cable, Quick Start Guide, Li-ion Rechargeable Battery, AC Adapter, Product USB, Brady Workstation Basic Design Software, Roll of M6-11-427 and M61-R4310 Ribbon
Languages Bulgarian, Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish, Swedish, Turkish
Memory 512 MB RAM, 256 MB Flash
Model M610
Print Speed 50.8 mm/sec.
Resolution 300 dpi
Connector Types USB 2.0
Weight (English) 3.1 lb.
Weight (Metric) 1.41 kg
Certifications/Compliance FCC Part 15 Class A
For Use With (Application) Asset Tracking, Circuit Board Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Safety Identification
Warranty 2 Years
Power Supply Details AC Adaptor, Rechargeable Battery
Voltage 110/240 V
Keyboard Type AZERTY
Alternate Part Number M610-AZERTY