Learn More
Alla France™ M50 Hydrometer
Product Code. 11743433
Change view
Click to view available options
Specific Gravity Range:
0.75 to 0.8 g/mL
0.8 to 0.85 g/mL
1.05 to 1.10 g/mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Specific Gravity Range unitSize
11743433 0.75 to 0.8 g/mL Each
11753433 0.8 to 0.85 g/mL Each
11723443 1.05 to 1.10 g/mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11743433 Supplier Alla France™ Supplier No. 3500MC075/20/Bqp
Alla France™ M50 Hydrometer is designed to measure the relative density of a liquid.
  • Calibration temperature 20°C
  • General purpose
Specifications
Type Hydrometer
Accuracy ±0.001
Increments 0.001 g/mL
Certifications/Compliance ISO 649-NF B 35511-BS 718-DIN 12791
Length (Metric) 270 mm
Specific Gravity Range 0.75 to 0.8 g/mL