Learn More
Alla France™ M50 Hydrometer
Click to view available options
Specific Gravity Range:
0.75 to 0.8 g/mL
0.8 to 0.85 g/mL
1.05 to 1.10 g/mL
Unit Size:
Each
Description
- Calibration temperature 20°C
- General purpose
Specifications
Specifications
| Type | Hydrometer |
| Accuracy | ±0.001 |
| Increments | 0.001 g/mL |
| Certifications/Compliance | ISO 649-NF B 35511-BS 718-DIN 12791 |
| Length (Metric) | 270 mm |
| Specific Gravity Range | 1.05 to 1.10 g/mL |