Learn More
Descrizione
Brady™ M210 Label Printers provides engineered, long lasting material quality with a two-year warranty. Extremely rugged and durable, the M210 Label Printer has been drop tested and is crafted with molded rubber bumpers to provide optimal protection from falls. This kit includes the M210 Printer, a lithium-ion battery, EU power cord, quick start guide, and label cartridge.
- Prints true-sized 6 to 19 mm wide identification to maximize amount of data
- Adjust length of labels using high-performance, continuous material
- Seven different industry specified materials that last
- Ergonomic cutter and label grabber that holds label after cutting
- Fast label creation with ABC keypad, graphics library, hot keys and menu functions
- Multi-functional accessory with magnet, flashlight and retractable printer stand for easy handling
- Smart Cell Technology for automatic label setup and formatting for wire wraps, terminal blocks, patch panels, cable flags and general labels
Specifica
Specifica
| Product Type | Label Printer |
| Display Type | LCD |
| Print Technology | Thermal Transfer |
| Width (English) | 4.5 in. |
| Width (Metric) | 114 mm |
| Height (English) | 9.5 in. |
| Height (Metric) | 241 mm |
| Depth (English) | 2.5 in. |
| Depth (Metric) | 64 mm |
| Color Capability | Monochrome |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.