Learn More
Brady™ M210 Label Printer - Carton Pack
Product Code. 17653215 Shop All Brady Products
Change view
Click to view available options
Color:
Yellow, Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17653215 Yellow, Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17653215 Supplier Brady™ Supplier No. 152260
  • ONLINE EXCLUSIVE PRICE
    117.00€
    110.00€ / Pack of 1
    Save 7.00€
    5% Off
View all promotions
 Restricted Item
In Stock
The M210 Label Printer is an all-in-one labeler for voice and datacom, electrical and industrial use, creating clear labels that stay stuck for years- despite extreme temperatures or highly textured surfaces.
Brady™ M210 Label Printers provides engineered, long lasting material quality with a two-year warranty. Extremely rugged and durable, the M210 Label Printer has been drop tested and is crafted with molded rubber bumpers to provide optimal protection from falls. This printer ships in a carton pack with a Quick Start Guide and an M21-750-499 label cartridge.
  • Prints true-sized 6 to 19 mm wide identification to maximize amount of data
  • Adjust length of labels using high-performance, continuous material
  • Seven different industry specified materials that last
  • Ergonomic cutter and label grabber that holds label after cutting
  • Fast label creation with ABC keypad, graphics library, hot keys and menu functions
  • Multi-functional accessory with magnet, flashlight and retractable printer stand for easy handling
  • Smart Cell Technology for automatic label setup and formatting for wire wraps, terminal blocks, patch panels, cable flags and general labels
  • Languages: Croatian, Czech, Danish, Dutch, English Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Specifications
Product Type Label Printer
Color Yellow, Black
Display Type Monocolor LCD
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Width (English) 4.5 in.
Width (Metric) 114 mm
Height (English) 9.5 in.
Height (Metric) 241 mm
Depth (English) 2.5 in.
Depth (Metric) 64 mm
Dimensions (L x W x H) 2.5 x 4.5 x 9.5 in.
Color Capability Monochrome
Includes Printer, Quick Start Guide and M21-750-499 Label Cartridge
Languages Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Memory 16 MB
Model M210
Print Speed 10 mm/s
Resolution 203 dpi
Weight (Metric) 0.75 kg
Certifications/Compliance 2011/65/EU and 2015/863
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, General Identification, Warehouse Marking
Warranty 2 Years
Power Supply Details AC Adaptor, Batteries, Rechargeable Battery
Voltage 110/240 V
Keyboard Type ABC
Alternate Part Number M210