Learn More
m-Tolylacetylene, 97%
Product Code. 11373986
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11373986
Brand: Thermo Scientific Alfa Aesar L17184.06
Request Bulk or Custom Format
CAS: 766-82-5 | C9H8 | 116.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 766-82-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD01318174
InChI Key RENYIDZOAFFNHC-UHFFFAOYSA-N
PubChem CID 136600
IUPAC Name 1-ethynyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C#C
Specifications
Density 0.9
Boiling Point 170°C to 175°C
Flash Point 50°C (122°F)
Odor Pungent
Refractive Index 1.544
Quantity 5 g
UN Number UN3295
Beilstein 2038007
Synonym 3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material m-Tolylacetylene