missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol-d7, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | m-Cresol-d7 |
| CAS | 202325-51-7 |
| Molecular Formula | C7 D7 H O |
| Quantity | 0.25 g |
| Synonym | 3-Methylphenol-d7, 1-Hydroxy-3-methylbenzene-d7, 3-Hydroxytoluene-d7, 3-Methylphenol-d7, NSC 8768-d7, m-Cresylic Acid-d7, m-Hydroxytoluene-d7, m-Kresol-d7, m-Methylphenol-d7, m-Oxytoluene-d7, m-Toluol-d7, meta-Cresol-d7 |
| InChI Formula | InChI=1 S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(O)c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 115.18 |
| Formula Weight | 115.1015 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction