Learn More
IKA™ M 20.1 Grinding Chamber
Shop All IKA Products
Click to view available options
For Use With (Equipment):
M20 Mill
Unit Size:
Each
Description
- The grinding chambers can be placed on the drive alternately
- One chamber is cleaned and filled while the other is being processed
Specifications
Specifications
| Product Type | Grinding Chamber |
| For Use With (Equipment) | M20 Mill |