Learn More
R&D Systems™ Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody Blocking Peptide
Product Code. 18268310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18268310 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18268310 Supplier R&D Systems™ Supplier No. NBP106640PEP
  • ONLINE EXCLUSIVE PRICE
    165.94€
    140.70€ / 0.05mg
    Save 25.24€
    15% Off
 Restricted Item
Estimated Shipment: 20-11-2026
A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A peptide to KDM6B. The Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 23135
Species Human
Product Line Novus
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-06640. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6 B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6 B, Lysine demethylase 6 B
Gene Symbol KDM6B
Product Type Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3
Quantity 0.05 mg