Learn More
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly
Product Code. 10053612 Shop All Labconco Corporation Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10053612 750 mL Each
10335130 500 mL Each
10734131 250 mL Each
10395170 1000 mL Each
10417400 100 mL Each
10318559 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10053612 Supplier Labconco™ Supplier No. 7554900
  • ONLINE EXCLUSIVE PRICE
    600.00€
    571.21€ / Pack of 1
    Save 28.79€
    4% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Complete three-piece flask consists of high-strength borosilicate glass molded top and bottom with silicone rubber seal
  • With 24/40 joints
  • Seal grasps top and bottom securely—never comes in contact with sample
  • Flat bottomed
  • Wide mouth lets you load samples easily and lyophilize quickly
  • Compatible with Labconco, Virtis™ and other major brands of freeze dryers
  • Flasks should be filled to no more than one-third the volume so that maximum surface area is achieved for efficient lyophilization
Required Accessories
  • Adapters are required to attach flasks to freeze dry valve ports
Specifications
Diameter (English) Outer 3.55 in.
Capacity (Metric) 750 mL
Material Borosilicate Glass
Height (English) 5.7 in.
Height (Metric) 14.5 cm
Includes Complete three-piece flask consists of high-strength Borosilicate Glass molded top and bottom with Silicone Rubber seal
Product Type Flask Kit
Standard Taper Joint 24/40
Weight (English) 3 lb.
Disposable Reusable
Mouth Style Joint
Bottom Shape Flat
Diameter (Metric) Outer 90.2 mm
Weight (Metric) 1.4 kg