Learn More
BUCHI Lyovapor™ L-250 Freeze Dryer
Shop All Buchi Products
Click to view available options
Style:
Not Software Compatible
Software Compatible
Unit Size:
Each
Description
- -85 °C condenser suitable for aqueous and organic-based solvent mixtures
- 4.5 L condensing capacity for standard laboratory applications
- EcoStream technology uses natural coolants
- Global Warming Potential (GWP) of only 4
- Reduced heat output and noise emissions in the lab
- Infinite-control technology offers process control via display, software, and mobile app
- Sample protection mode prevents sample collapse
Specifications
Specifications
| Type | Benchtop Freeze Dryer |
| Capacity (Metric) | 4.5 L |
| No. of Shelves | 0 |
| Temperature Range (Metric) | Ambient to -105°C |
| Material | Stainless Steel Coil and Collector Chamber |
| Style | Not Software Compatible |
| Certifications/Compliance | Low Environmental Impact Factor |
| Plug Type | All except North America |
| Voltage | 220/240 V |