Learn More
STEMCELL Technologies Lymphoprep™
Product Code. 15257179
Encompass_Preferred
Change view
Click to view available options
Product Type:
250 mL
1000 mL
3000 mL
Quantity:
1 x 250 mL Bottle
4 x 250 mL Bottle
6 x 500 mL Bottles
Unit Size:
250mL
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
15257179 3000 mL 6 x 500 mL Bottles Pack of 6
17171036 250 mL 1 x 250 mL Bottle 250mL
17181036 1000 mL 4 x 250 mL Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15257179 Supplier STEMCELL Technologies Supplier No. 07861
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Density gradient medium for the isolation of mononuclear cells
  • Lymphoprep™ is a density gradient medium recommended for the isolation of mononuclear cells from peripheral blood, cord blood, and bone marrow by exploiting differences in cell density
  • Granulocytes and erythrocytes have a higher density than mononuclear cells and therefore sediment through the Lymphoprep™ layer during centrifugation
  • Lymphoprep™ can be substituted for Ficoll-Paque™ without any need to change your existing protocols and is fully compatible with both SepMate™ and RosetteSep™
  • Lymphoprep™ has a density of 1.077 g/mL
Specifications
Quantity 6 x 500 mL Bottles
Cell Type Mononuclear Cells
For Use With (Application) Cell Isolation
Product Type 3000 mL