missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat anti-Mouse, PE, Clone: 1A8, BD
Rat Monoclonal Antibody
143.00€ - 456.00€
Specifications
Antigen | Ly-6G |
---|---|
Clone | 1A8 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15810779
|
BD Biosciences
561104 |
25 μg |
143.00€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
15886658
|
BD Biosciences
551461 |
0.2 mg |
456.00€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The 1A8 monoclonal antibody specifically binds to Ly-6G, a 21-25-kDa glycosyolphosphatidylinositol-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The RB6-8C5 monoclonal antibody recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G.
Specifications
Ly-6G | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Ly-6G-transfected EL4J cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
1A8 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1 |
For Research Use Only.