Learn More
(-)-Lupinine, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Lupinine |
| CAS | 486-70-4 |
| Molecular Formula | C10 H19 N O |
| Quantity | 50 mg |
| Synonym | (R,R)-1-(Hydroxymethyl)octahydroindolizine |
| InChI Formula | InChI=1S/C10H19NO/c12-8-9-4-3-7-11-6-2-1-5-10(9)11/h9-10,12H,1-8H2/t9-,10+/m0/s1 |
| SMILES | OC[C@@H]1CCCN2CCCC[C@H]12 |
| IUPAC Name | [(1R,9aR)-2,3,4,6,7,8,9,9a-octahydro-1H-quinolizin-1-yl]methanol |
| Molecular Weight (g/mol) | 169.26 |
| Formula Weight | 169.1467 |