Learn More
Hamilton™ Luer to Small Hub RN Adaptor
Product Code. 12376239
Change view
Click to view available options
Product Type:
Adapter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12376239 Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12376239 Supplier Hamilton™ Supplier No. 5575301
  • ONLINE EXCLUSIVE PRICE
    69.00€
    62.29€ / Pack of 1
    Save 6.71€
    9% Off
Estimated Shipment: 14-12-2026
Small Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 100 μL and smaller. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Specifications
Product Type Adapter