Learn More
Value Plastics Luer Adapter Fittings, Tee, Polypropylene
Shop All Cole-Parmer Products
Click to view available options
Width (English):
29/32 in.
37/64 in.
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards. Constructed of ADCF polypropylene
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer Thread, Male Slip Luer |
| Width (English) | 29/32 in. |