Learn More
R&D Systems™ LTB4 Parameter Assay Kit
Product Code. p-4753769
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
Product Code. 18762064
Brand: R&D Systems™ KGE006B
LTB4 Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The LTB4 Parameter Assay Kit quantifies multi-species Leukotriene B4/LTB4 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 10.30 to 2500 pg/mL
Assay Sensitivity 10.9 pg/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity LTB4 Levels in Various Samples
Sufficient For 1 x 96-well Plate
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.