missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LSN 2463359

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
LSN 2463359 is a potent and selective positive allosteric modulator at the mGlu5 receptor (EC50 = 24 nM). Has no activity at mGlu1-4, mGlu8 and GABAB receptors. Reverses methylazoxymethanol acetate (MAM)-induced deficits in reversal learning memory and normalizes SDZ 220-581 and RO 63-1908 -induced alterations in instrumental responses in rat models. Promotes wakefulness in vivo. Brain penetrant and orally active.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | LSN 2463359 |
| Purity | >99% |
| Molecular Weight (g/mol) | 265.31 |
| Inhibitors | LSN 2463359 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction